Biotix Assay Plates, Biotix
Supplier: Avantor
Certificates
About this item
The large well diameter of Biotix 96-Well assay plates make them ideal for a broad range of applications, including compound libraries and combinatorial chemistry
Made of non-treated, medical-grade virgin polypropylene, these 96-Well plates conform to the ANSI/SBS 1-2004 standard for automated high throughput processing. The raised "chimney" wells reduce cross-contamination and allow secure application of cap mats and sealing films. An alphanumeric grid helps with sample identification and a notched bottom left corner ensures correct plate orientation. Select either V-shaped or U-shaped well bottom plates for specified assay needs.
Customers Who Bought This Also Bought