SP Bel-Art Standard Stem Funnels, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Polypropylene plastic funnels feature an exclusive filter-step internal design for speedier filtrations.
- Polypropylene plastic construction
- External ribs
- Autoclavable
- Filter-step internal design
External ribs prevent airlock. The parallel stem measures 8 mm O.D. Steam autoclavable at 121ºC (250ºF).
Customers Who Bought This Also Bought