Glass Precision Ground Button Type Stirrer Shaft, 19 mm, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
The 19 mm O.D
button type with a hollow shaft is for use with all 19 mm I.D. ACE Trubore® bearings. Also compatible with a mechanical seal and PTFE bearings. Fits certain ACE chucks. Ground surface approximately 380 mm long.
Customers Who Bought This Also Bought