Large Scale Spherical Reaction Flask Support Stand, Ace Glass Incorporated
Catalog # 89054-872
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacity:200 L
- Description:Large Scale Spherical Reaction Flask Support Stand
- Cat. no.:89054-872
- W×D×H:42 x 42 x 90"
- Supplier No.:6474-373