ContactUsLinkComponent
Unjacketed, Spherical Pilot Plant Reactor System, 200 L, Ace Glass Incorporated
Unjacketed, Spherical Pilot Plant Reactor System, 200 L, Ace Glass Incorporated
# de catalogue 89054-880
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
6474 Pilot Plant Reactor, Spherical, 50L, 72L, 100L, 200L
Unjacketed, Spherical Pilot Plant Reactor System, 200 L, Ace Glass Incorporated
# de catalogue 89054-880
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Complete Unjacketed Spherical Pilot Plant Reactor System, KF Plane Flange
  • Cat. no.:
    89054-880
  • Supplier No.:
    6474-75
Spécifications
A propos de cet article
Complete cylindrical pilot plant reactor on self-supporting, mobile support frame with 1" stainless steel pipe
Flask opening is 300 mm (12") flat-ground flange. Bottom outlet is a flush seal drain valve with one-inch beaded drain discharge. O-Ring supplied on valve is a CAPFE, silicone encapsulated with PTFE, and Kalrez for excellent chemical resistance. Flask has 100 mm I.D. side port, angled 45 degrees. Motor is secured on black epoxy coated mounting plate over center standard taper 45/50 opening on head; other openings are for condenser, thermoprobe, cooling coil, etc. Head is ­secured to flask with PTFE gasket and plastic coupling. Maximum bolt tightening torque for coupling (cold state) is 30 in lbs. Stirring motor is heavy duty, 1/2 HP electric motor for hazardous area duty. Motor shaft is 5/8" O.D. Controller operates on 120V, 50/60 Hz.
Features a 28 mm (1 1/8") O.D. precision ground and polished glass stirring shaft with a "pinned" multi-paddle PTFE agitator at bottom and a turbine blade PTFE agitator higher on shaft for grease-free operation with PTFE bearing. Cooling coil is 1/2" O.D. copper tubing encapsulated with PTFE; also available in 316 stainless steel or Hastalloy C-276. A digital temperature control system is supplied to control heating mantles. This prevents over-heating and overshoot, and gives an instant readout of temperature.
Measures 90" high, 42" wide, 42" deep. Note: Frame shipped assembled. Black epoxy coated aluminum flask support plate is adjustable, vertically, to allow room under flask for various outlet configurations. Upper support brackets supplied to stabilize flask. Head is traditional domed style, with seven openings; five standard taper 45/50, two standard taper 29/42.
Customers Who Bought This Also Bought