Spécifications
- Capacity:750 ml
- Dimensions:18 x 11"
- Description:Benchtop Absorbent Mat
- Cat. no.:89170-938
- Supplier No.:10324
Spécifications
A propos de cet article
Aqui-Pad™ is specifically designed for use on the laboratory benchtop to absorb spills or drops of aqueous liquid during routine lab work
Aqui-Pad™ consists of two material layers with absorbent media sealed inside. The top layer is a non-woven polypropylene pad and the bottom layer is impervious polypropylene film to enclose any liquid spills so the liquid does not reach the benchtop. Inside the two layers is a proprietary absorbent media fabricated from tissue paper and wood pulp materials that are impregnated with a super absorbent polymer. Aqui-Pad™ is supplied in flat sheets 18" x 11" (457mm x 280mm) and will absorb up to 750 mL. Place in biohazard disposal after use.