A propos de cet article
Absorbent sheets are the most cost-effective absorbent product that complies with the regulations governing specimen transport.
- Tissue-based
- Impregnated with super-absorbent polymers to absorb liquids that may escape during transport
Sheets can be purchased in a variety of stock sizes or custom cut to meet your specific requirements.