Glas-Col® Aluminum Housing Heating Mantle, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
This mantle comes with a hole in bottom to accommodate flasks with bottom outlets
It offers the benefit of grounding through aluminum housing. The heating element is embedded in layers of glass fabric to protect the flask wall from thermal strain, and is thoroughly covered with glass insulation to prevent heat from being radiated outward. Glass fabric affords operating temperatures to 450°C. With detachable 4', 3-wire cord and locking connector. All 115V Mantles are CSA Certified.
Customers Who Bought This Also Bought