Single Blade PTFE Rotating Agitator, PTFE, 19 mm, Ace Glass
Supplier: Ace Glass
Certificates
About this item
PTFE agitator with removable blade secured with a PTFE pin
For use with 19 mm Polished Shafts, Precision Ground Shafts, and PTFE Covered Stainless Steel Shafts.
Blade can be rotated to fit through a standard taper 45/50 joint opening. Fits 19 mm od drilled or knob shafts.
Customers Who Bought This Also Bought