Clamp Lamp with Reflector, Medium Pressure, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Reflector, for use with Ace-Hanovia U.V
lamps. Reflects 85% of spectral rays. Made of aluminum with 44.5 x 10 cm opening. Lamp ends are adapted for reflector. Holes in reflector are drilled to accept 11.4 cm lamp and are adjustable for 19.1 and 30.5 cm lamps. Lamp's arc length is 11.4 cm, 4.5". Larger lamps available on special order. Reflector supplied with clamps on rear brackets for mounting to 1/2" rod not supplied and 6 ft power cord. Complete unit consists of reflector, lamp and power supply. All parts must be ordered separately.
Customers Who Bought This Also Bought