Multi-Paddle Agitator, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
For use with a stainless steel shaft or a borosilicate straight shaft
Center hole has a 10 mm inner diameter. and one or more can be positioned on shaft at various locations using set screws to secure. With stainless shaft use stainless set screw; with glass shaft use Kel-F-tipped set screw. Both screws supplied with agitator.
Customers Who Bought This Also Bought