Covered Stainless Steel Stirrer Shaft, PTFE, 19 mm, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Precision PTFE covered stainless steel stirring shafts, 19 mm O.D
Drilled type shafts accept agitators and can be used in 19 mm Trubore® bearings. Join to stirrer motor using certain ACE series chucks.
Customers Who Bought This Also Bought