Select Shoulder Split Leather Palm Gloves with 4" Gauntlet Cuff, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
These gloves feature selected shoulder split leather with striped canvas back and cuff.
- Knit wrist
- Full leather palm and fingertips
- Wing thumb
A cuff and knit wrist prevent debris from entering the glove and irritating skin.
Specifications
- Catalog No:
- 80092-086
- Ambidextrous/Hand-specific:
- Hand-specific
- Disposable / reusable:
- Reusable
- Cuff style:
- Gauntlet
- Cuff length:
- 4"