Grain Cowhide Welder's Gloves, Wells Lamont
Fournisseur: Wells Lamont
Certificats
A propos de cet article
These gloves provide increased resistance to high heat.
- Wing thumb
- Ambidextrous
Featuring quality grain cowhide with split leather 4" cuff, these gloves have natural heat and abrasion resistance with good tensile strength.
Spécifications
- Catalog No:
- 80092-034
- 80092-038
- 80092-036
- 80092-040
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous