Scepter™ Sterile M321 Glove Liner, Wells Lamont
Fournisseur: Wells Lamont
Certificats
A propos de cet article
This sterile, cut-resistant liner is made of thin-gauge stainless steel wrapped in polyester.
- Ambidextrous
It can be washed 15 times without shrinking.
Certifications: ANSI Cut Level 4.
Ordering information: Glove Liners are sold 20 by box.
Spécifications
- Catalog No:
- 52428-890
- 52428-894
- 52428-896
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous