SP Bel-Art Spinstar® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Spinstar® magnetic stirring bars create a deep mixing vortex at relatively slow speeds.
- Perfect for applications requiring slow, thorough mixing
- PTFE low-friction coating
- Durable and inert
- Autoclavable
Star shape is designed to fit the inside diameter of most commonly-used beakers.
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought