SP Bel-Art Spinbar® Color Octagon Magnetic Stirring Bars, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
- PTFE low-friction coating
- Durable and inert
- Interrupted profile
- Pivot ring aids in reducing friction and chattering
- Autoclavable
Interrupted profile provides greater surface area and added turbulence
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Choose from red, yellow or blue.
Customers Who Bought This Also Bought