Spécifications
- Description:PBS Tablets Sufficient for 100 mL
- Size:100 Tablets
- Cat. no.:89284-786
- Supplier No.:GTX48887
Spécifications
A propos de cet article
Pre-measured Phosphate Buffered Saline Tablets
Each tablet prepares a 1X PBS solution when dissolved in 100 mL of ddH2O. For in vitro research use only. Not intended for any diagnostic or therapeutic purpose. Not suitable for human or animal consumption. Store at room temperature.