SP Bel-Art Electrode Rack, Bel-Art Products, a part of SP
# de catalogue 34102-089
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Electrode Rack
- Cat. no.:34102-089
- Supplier No.:H18509-0000
Spécifications
A propos de cet article
Handy rack holds 8 test tubes for storing and conditioning pH and ion electrodes.
- Polypropylene construction
- Suitable for storing and conditioning electrodes
- Chemically resistant
- Autoclavable
The polypropylene rack resists chemical attack and is autoclavable at 121 °C (250 °F). Rack accomodates two rows of 4 tubes, up to 20 mm in diameter.
LxWxH: 10.8 x 6 x 12.1 cm
Customers Who Bought This Also Bought