SP Bel-Art Spinbar® Jumbo Magnetic Retriever, Bel-Art Products, a part of SP
Catalogue # 47750-234
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Jumbo Stirring Bar Retriever
- Cat. no.:47750-234
- Supplier No.:377810000
Spécifications
À propos de cet article
This flexible PTFE magnetic retriever will remove the largest magnetic stirring bars from deep vessels.
- PTFE construction
- Easily remove magnetic stirring bars
- Easy-grip handle
Retriever has an easy grip “T” handle, with an overall length of 65 cm (26"). This includes the powerful Alnico magnetic end, which is 10.2 x 1 cm diameter (4 x 3/8").
Ordering information: Longer, custom lengths are available.