SP Bel-Art Magnetic Stirring Bar Arrester, Bel-Art Products, a part of SP
# de catalogue 13197-258
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Coating:Polyethylene
- Description:Stirring Bar Arrester
- Cat. no.:13197-258
- Supplier No.:F377860000
Spécifications
A propos de cet article
The Magnetic Stirring Bar Arrester holds your stirring bar in place when pouring from a beaker or flask by placing the arrester against the bottom of the container.
- Polyethylene construction
The arrester is 9.5 mm (3/8") thick polyethylene, 51 mm (2") diameter with a radial magnet.
Ordering information: Other sizes can be made to order.
Customers Who Bought This Also Bought