SP Bel-Art Spinpak® Magnetic Stirring Bar Six Pack Assortments, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These bars are best suited for specific needs.
- Choose from octagonal, round, micro, or color-coded bars
- PTFE low-friction coating
- Durable and inert
- Convenient assortment packs suit a variety of needs
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought