Spécifications
- Coating:PTFE
- Ø:53 mm (2")
- Length:13 mm (1/2")
- Cat. no.:89042-248
- Supplier No.:F371520000
Spécifications
A propos de cet article
The Capsule Magnetic Stirring Bar is a polystyrene capsule containing a 30 x 8 mm (1.18 x 0.31") polygon magnetic stirring bar that spins freely inside the capsule while being prevented from spinning off center and ‘wandering’ on the base of a vessel.
- Stirring bar contained to prevent damage to pH probes
- Will not interfere with gels during staining procedures
- Capsule can be easily disassembled for cleaning
- Autoclavable
Containment of the stirring bar prevents it from impacting and damaging expensive pH probes during pH adjustments, and from interfering and damaging gels during staining/destaining procedures.
Large, flat capsule keeps the stirring bar consistently centered on the bottom of vertical electrophoresis units (e.g. Hoefer’s) to efficiently mix the running buffer.
Use of the capsule prevents the stirring bar from being impeded by chemicals which, in water, transform into a sticky gel (e.g. albumin) and enables efficient dissolving.
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.