SP Bel-Art Spinplus® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Spinplus® magnetic stirring bars add speed and efficiency to mixing operations.
- The “+” shape creates a deep vortex and provides stable, quiet operation
- PTFE low-friction coating
- Durable and inert
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought