Color Frosted Beveled Edge Microscope Slides, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
Globe Scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
- Dimensions: 25×75×1 mm
- Thickness: 1.1 mm
- Color frosted one end, one side
These color frosted microscope slides are offered in bright, attractive colors. They feature beveled edges with clipped corners. The slides are pre-cleaned and supplied ready for use.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).
Customers Who Bought This Also Bought