ContactUsLinkComponent
EZ-Rate™ Westergren Pipettes, Globe Scientific
EZ-Rate™ Westergren Pipettes, Globe Scientific
# de catalogue 10027-470
Fournisseur:  Globe Scientific
Westergren ESR Pipette (For Direct Testing In 13Mm Blood Collection Tubes),200 Tests/Box
EZ-Rate™ Westergren Pipettes, Globe Scientific
# de catalogue 10027-470
Fournisseur:  Globe Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EZ-Rate™ Westergren Pipettes
  • Cat. no.:
    10027-470
  • Supplier No.:
    3480
Spécifications
A propos de cet article
The EZ-Rate™ offers a simple, safe, economical and highly-accurate method for performing Westergren ESR determinations directly in 13 mm sodium citrate blood collection tubes.
The polystyrene EZ-Rate™ pipette is graduated from 0 to 180 mm and features a unique fibrous plug located at the zero mark. The plug is specifically designed to be a self-sealing aerosol barrier that stops hazardous substances from escaping through the top of the pipette.
Customers Who Bought This Also Bought