Autoclavable Water Bottle, Tecniplast®
# de catalogue 10717-475
Fournisseur: Tecniplast Usa
Product available on GSA Advantage®
Spécifications
- Capacity:150 ml (5 oz.)
- Description:Water Bottle, without Caps
- Material:Polycarbonate
- Cat. no.:10717-475
- Supplier No.:ACBT0150
Spécifications
A propos de cet article
Shatterproof, transparent, autoclavable water bottle with permanent graduations.
Bottles fit under filter-style covers. Designed for cage sizes A and B.
Customers Who Bought This Also Bought