B/T Sure™ Biological Indicators, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Indicators verify the sterility of instruments, vials, glassware, and other products in less time than it takes to send samples to an outside lab.
- Use to verify sterility
- Purple color ensures sterilization
Purple color ensures sterilization. Color change from purple to yellow indicates an ineffective sterilization process. Indicators are supplied in a variety of packaging options, with each vial containing a spore population of either 105 or 106.
Customers Who Bought This Also Bought