Sampling Thief Tube, Ace Glass Incorporated
Catalog # 89063-440
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Ext.Ø:1"
- Description:Sampling thief tube
- Length:1016 mm (40")
- Cat. no.:89063-440
- Supplier No.:8664-05
Specifications
About this item
Used to take samples from large vessels such as 55 gallon drums.
Measures 40L×1" Ext.Ø with tooled tip at bottom and bulb handle at top for secure grip, open at both ends.
Customers Who Bought This Also Bought