Tilting Support, Glas-Col, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This is a tilting support that provides a finger tip method of emptying flasks by pouring
Mantle support ring may be adjusted to and locked at any convenient height. Upper cross member strengthens assembly; serves as stop for tilted mantle. Base is compact, 91 x 98 cm, but extremely stable. Complete item sold with one set of rings.
Customers Who Bought This Also Bought