Tripod Support, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
Certificats
A propos de cet article
For 3,000mL- 72,000mL Glas-Col Mantles
The flask capacity 12000-72000 mL supports contain an adjustable bracket to adjust height between 24 to 36" in 3" increments.
Customers Who Bought This Also Bought
VWR® Volumetric Flask, Clear Glass, Narrow Neck, Class A, Unserialized
Fournisseur: VWR International
$342.28 to $622.36
VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Fournisseur: VWR International
$615.51 to $938.06
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Kimble Chase
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
$357.82 to $2,899.60
VWR® Graduated Cylinders, Calibrated To Contain, Class B
Fournisseur: VWR International
$79.45 to $714.70
VWR® Volumetric Pipettes, Class A, Serialized, Color-Coded
Fournisseur: VWR International
$127.13 to $508.47
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialized
Fournisseur: VWR International
$127.29 to $1,375.41
VWR® Volumetric Flask, Clear Glass, Wide Neck, Class A, Unserialized
Fournisseur: VWR International
$270.79 to $738.55
VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Serialized
Fournisseur: VWR International
$42.90 to $467.60
SP Wilmad-LabGlass Volumetric Color Coded Class A Pipettes, SP Industries
Fournisseur: SP Industries
$151.42
PYREX® Economy Double Metric Scale Cylinders, To Contain, Corning
Fournisseur: Corning
$60.30 to $716.80
Hydrometer Cylinders, Borosilicate Glass, United Scientific Supplies
Fournisseur: United Scientific Supplies
$99.30
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 One Mark, Bulb Pipette, with Batch Certificate icate 3.3 USP Type I, 1.5 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 1.5 mL, Class/ Quality Grade: Type I, Class A, Tolerance: 0.006 mL,
Fournisseur: Foxx Life Sciences
$194.12
PYREX® Dispensing Burets, Colored Scale, with Straight Bore PTFE Stopcock, Corning
Fournisseur: Corning
$716.77