Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
Certificats
A propos de cet article
Synonymes: Methyl cyanide,Ethanoic acid nitrile,Cyanomethane
Formule:H₃CCN
MW:41.05 g/mol
Point d'ébullition:81,6 °C (1013 hPa)
Point de Fusion:-45,7 °C
Densité:0,782 g/cm³ (20 °C)
Numéro MDL:MFCD00001878
Numéro CAS:75-05-8
Index Merck:13,00071
:Acétonitrile
:Storage Temperature: Ambient
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Point d'éclair:2 °C (closed cup)
:Point d'ébullition:
:Pt ébul.: 81,6 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: -45,7 °C
:Point eclaire:
:Pt Eclair: 2 °C (coupe fermée)
:M.W. 41,05 g/mol
EINECS:200-835-2
:-45,7
:81,6
ONU:1648
:01-2119471307-38
:3
:II
:2
:GHS02
:GHS07
Température d'entreposage:Température ambiante
:Filter 0,2 um_Nitrogen_filled
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Résultats des tests de spécification
- Assay:Min. 99.9 %
- Solution (100 g/L):Neutral
- Acidity:Max. 0.0008 meq/g
- Alkalinity:Max. 0.0006 meq/g
- Coloration:Max. 10 APHA
- n 20/D:1.343 to 1.345
- Evaporation Residue:Max. 2 ppm
- Water:Max. 30 ppm
- Absorbance (200 nm):Max. 0.10
- Transmittance (190 nm):Min. 10 %
- Transmittance (195 nm):Min. 80 %
- Transmittance (200 nm):Min. 95 %
- Transmittance (210 nm):Min. 96 %
- Transmittance (220 nm):Min. 97 %
- Transmittance (230 nm):Min. 98 %
- Transmittance (240 nm):Min. 99 %
- Transmittance (250 nm):Min. 99 %
- Transmittance (240 to 420 nm):Min. 99 %
- Fluorescence (as quinine) (254 nm):Max. 1 ppb
- Gradient grade (210 nm):Max. 10 mAU
- Conforms to ACS:Passes Test
- Conforms to ACS (HPLC/UV):Passes Test
- Conforms to Reag. Ph.Eur.:Passes Test
- Conforms to USP:Passes Test
- VWR Chemicals BDH®, Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC: