Hyvac Light Pump Oil, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Hyvac Light Pump Oil incorporates two essential high vacuum characteristics: low vapor pressure which does not materially increase at temperatures up to 50 °C, and viscosity sufficiently low for use at 15 °C
It remains fairly constant up to 50 °C. For use with belt-driven or direct-drive pumps.
Customers Who Bought This Also Bought