9 mm Polished Glass Stirrer Shaft, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Polished, 9mm OD shaft for use in liquid seal bearings and ACE debris free stirrer assemblies
Shaft length accommodates flasks from 500 to 5000 mL.
Customers Who Bought This Also Bought