Melting Point Tubes, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Tubes are manufactured from borosilicate glass and packaged 100 tubes per vial. Closed at one end.
Tube 80061-554 has a smaller I.D. to conform to U.S.P XXI requirements.
Customers Who Bought This Also Bought