Tripod Support Stand for 100 and 150 mm Columns, Ace Glass Incorporated
# de catalogue 89076-384
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Tripod Support Stand
- Height:14"
- Cat. no.:89076-384
- Supplier No.:12099-20
Spécifications
A propos de cet article
Tripod support for bottom of Ion-exchange columns
Metal support has hole large enough for end fitting to pass through so that shoulder of column rests on plate. Column should also be stabilized with a clamp. For use only with 100 and 150 mm diameter columns.
Customers Who Bought This Also Bought