Support Tripod, Glas-Col, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This tripod is used for 5 L, 12 L, and 22 L heating mantles with a soft glass outer covering instead of spun aluminum.
Customers Who Bought This Also Bought