Extension Support, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Provides convenient supports for flasks up to five-liter capacity
Generally used with flasks having aluminum housing.
Customers Who Bought This Also Bought