Labjaws FootPlate, Ace Glass Incorporated
# de catalogue 89084-716
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Labjaws FootPlate for Frame
- Cat. no.:89084-716
- Supplier No.:11174-13
Spécifications
A propos de cet article
The Labjaws Footplate holds Flexaframe poles
It secures them to floors, walls and tables with screws. Mount lattice permanently.
Customers Who Bought This Also Bought