Tube, Break Seal, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
Certificats
A propos de cet article
Used as a one-time opening valve
Drawn to a point so that a small weight will fracture seal.
Customers Who Bought This Also Bought
Economy Certified Glass VOA Vials with 0.125" Septa, Thermo Scientific
Fournisseur: Thermo Scientific
$382.27
Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$1,001.33
I-CHEM™ Vials with Screw Cap, Borosilicate Glass, Thermo Scientific
Fournisseur: Thermo Scientific
$563.12
KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$105.21