Power Outlet Strip, Ace Glass
# de catalogue 89076-710
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Power Outlet Strip, Four Outlets, NEMA 5-15P
- Spacing:1.25" (center to center)
- Cat. no.:89076-710
- Electrical:120 VAC, 50/60 Hz, 15 A
- Cord Length:6'
- Output:(4) NEMA 5-15R
- Supplier No.:12195-20
Spécifications
A propos de cet article
This power outlet strip has four outlets
Furnished with 15 amp push-to-reset circuit breaker, U‑ground outlets with electrically wired ground and heavy duty UL listed cord set. Sturdy, blue finished steel housing. Rated 15 amps, 130 volts continuous duty. Shipping weight, 2 lbs.
Customers Who Bought This Also Bought