Flat Flange Reaction Head, 4 Neck, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
For use with ground flat flange flasks
With three in-line standard taper joints and a standard taper 10/30 side joint for thermometer. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought