Adapter, Twin Hose Connection, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This adapter is normally used with a reflux apparatus to allow inert gas flow over top of the apparatus to maintain an oxygen free system.
- [ST] inner joint
- Twin hose connections opposite to each other
- Fabricated from borosilicate glass
This adapter has a standard taper inner joint and twin hose connections opposite to each other. Use with a 3/8" or 5/16" inner diameter tubing, size C hose connection.
Customers Who Bought This Also Bought