Microscope Slide Storage Boxes for 12 Slides, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
Globe's microscope slide storage boxes are designed for the safe and long term storage of microscope slides
An inventory sheet is located on the inside cover for convenient slide identification and organization. The base of the box is cork lined for added protection.
They are produced from durable ABS plastic and offer a hinged lid for easy opening and closing.
Ordering information: Available in 4 sizes, these storage boxes accommodate 25 x 75 mm and 3 x 1" microscope slides.
Customers Who Bought This Also Bought