VWR® Vacuum Ovens
Fournisseur: VWR International
Certificats
À propos de cet article
The 12.5 L unit features a top-mounted independent evacuation and venting vacuum fittings.
- Heating system optimizes chamber space for drying, curing, vacuum embedding, and plating applications
- Polycarbonate safety shield protects door glass
- Built-in over-temperature protection
- Vacuum level is displayed on gauges from 0 to 30 in Hg
- Corrosion-resistant stainless-steel tubing for long-term performance
- With two removable stacking aluminum shelves
- Easy-to-clean Type 304 stainless-steel interior chamber
- Powder-coated heavy-gauge steel exterior for durability
- Two-year warranty with extended warranty packages available
The 19.8 L and 65.1 L units feature a front-mounted three-way valve for evacuation, venting and purging of inert gases (e.g., Nitrogen or Argon) with vacuum fittings on the front of the unit.
7.6 cm (3") glass wool insulation prevents heat loss. Silicone door gasket and positive latch door maintain seal at all vacuum levels.
Ordering information: Comes with Two-Year Parts and Labor Warranty.
Informations connexes
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only). Call 888-793-2300 for more details. Extended warranty plan extends the equipment's original warranty to provide better coverage for your equipment. Each extended warranty is ONLY valid at the point of sale. Extended warranty applies for US only. Each extended warranty includes unlimited labor, parts, and travel as well as priority phone and e-mail support. Service performed by experienced qualified technicians.
White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer's receiving dock.
White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer's receiving dock.
Frequently Bought Together