VWR® Mechanical Pump Oil No. 21
# de catalogue 40300-028
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Description:VWR® Mechanical Pump Oil No. 21
- Size:1 L (33.8 oz.)
- Cat. no.:40300-028
Spécifications
A propos de cet article
Designed for use in applications where reactive or corrosive gases are present.
- Resistant to oxidation and decomposition
- Good vapor pressure capabilities
Additives resist oxidation and decomposition. The oil uses molecular distillation to ensure good vapor pressure capabilities. Vapor Pressure at 25 °C is 3e-5torr. Boiling Point at 0.01torr is 112 °C.
Customers Who Bought This Also Bought