Titration Cup, Dyn-A-Med
# de catalogue 12578-108
Fournisseur: Dyn-A-Med
Spécifications
- Capacity:100 ml (3.4 oz.)
- Description:Titration Cup
- Cat. no.:12578-108
- Supplier No.:80099
Spécifications
A propos de cet article
Made of polypropylene.
Graduated on two sides and has a writing surface. For METTLER TOLEDO® DL series titrators.
Customers Who Bought This Also Bought