Water-Cooled Bearing Trubore® Tubing, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Interchangeable, ground joint bearing with jacket for water cooling.
May be operated as high as 2000 rpm using ACE high-speed lubricant. Can be operated at 1500 rpm unevacuated using ACE Stir-Lube. Hose connections for use with 5/16" or 3/8" Int.Ø tubing. Available with PTFE-clad joint to eliminate the need for grease. Use with ACE precision ground stirring shafts.
Customers Who Bought This Also Bought