Bearing, High Speed Vacuum, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Used with aluminum packing box
For shaft speeds of 1000 r.p.m. and higher, and vacuum operation down to 0.5 mm Hg. The seal is made entirely by the packing box. Only infrequent lubrication is required using packing and Stir-Lube. We recommend that you use certain ACE vacuum adapters to avoid contaminating the flask contents with lubricant. For use with 9 mm O.D. shafts.
Customers Who Bought This Also Bought