Krytox LVP High Vacuum Grease, Ace Glass Incorporated
Catalogue # 89062-222
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Temperature Range:–20…+260 °C (–5…+500 °F)
- Description:Krytox LVP High Vacuum Grease
- Size:2oz.
- Cat. no.:89062-222
- Supplier No.:8116-10
Spécifications
À propos de cet article
This grease has very low a vapor pressure, highly inert, and nonflammable
Krytox LVP high vacuum grease is a combination of an extremely low vapor pressure perfluoroalkylpolyether oil and a fluorocarbon resin thickener. This white, buttery grease is designed to lubricate the fittings and accessories of high vacuum systems at operating temperatures down to 10-12 torr at 20°C (1.33 x 10-13 kPa).
The grease is for high-vacuum systems. Superior performance in laboratory and pilot plant equipment, as a lubricant and sealant for stopcocks, valves, fittings and o-rings operating at high vacuum or in hostile environments.